Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alpelisib ≥99%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A130937-100MG A130937-1MG A130937-250MG A130937-50MG A130937-5MG A130937-10MG
77009-938EA 313 USD InStock
77009-938 77009-940 77009-942 77009-944 77009-946 77683-164
Alpelisib ≥99%, Ambeed.Inc
Alpelisib
Precaución: For research use only.


Formula: C₁₉H₂₂F₃N₅O₂S
MW: 441.47 g/mol
MDL Number: MFCD22417085
CAS Number: 1217486-61-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty