Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Ethoxymethyl)furan 98%

Supplier: AMBEED, INC
Warning

A131375-100G A131375-10G A131375-1G A131375-25G A131375-5G
76983-720EA 690 USD InStock
76983-720 76983-722 76983-724 76983-726 76983-728
2-(Ethoxymethyl)furan 98%
2-(Ethoxymethyl)furan
Precaución: for Research Use Only


Formula: C₇H₁₀O₂
MDL Number: MFCD02258625
CAS Number: 6270-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty