Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium bis(2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate) 96%

Supplier: AMBEED, INC
Warning

A131421-100G A131421-25G A131421-500G A131421-5G
77027-076EA 66 USD InStock
77027-076 77027-078 77027-080 77027-082
Sodium bis(2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate) 96%
Sodium bis(2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate)
Precaución: for Research Use Only


Formula: C₁₂H₂₂O₂
MDL Number: MFCD02094609
CAS Number: 103404-87-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty