Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxycinnamaldehyde 97%

Supplier: AMBEED, INC
Warning

A132061-1G A132061-25G A132061-5G
76983-770EA 15 USD InStock
76983-770 76983-772 76983-774
4-Methoxycinnamaldehyde 97%
4-Methoxycinnamaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD00017343
CAS Number: 1963-36-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty