Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%

Supplier: AMBEED, INC
Warning

A132635-250MG A132635-5G
76996-466EA 8 USD InStock
76996-466 77566-572
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine
Precaución: For research use only.


Formula: C₃₀H₃₀N₂O₇
MDL Number: MFCD00063410
CAS Number: 23669-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty