Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one 98%

Supplier: AMBEED, INC
Warning

A133084-100G A133084-25G A133084-5G
76996-492EA 82 USD InStock
76996-492 76996-494 76996-496
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one 98%
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one
Precaución: for Research Use Only


Formula: C₁₄H₁₈N₂O₅
MDL Number: MFCD02678087
CAS Number: 179688-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty