Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-2-fluoroaniline 97%

Supplier: AMBEED, INC
Warning

A133245-100MG A133245-1G A133245-250MG A133245-5G
77027-158EA 20 USD InStock
77027-158 77027-160 77027-162 77027-164
5-(Benzyloxy)-2-fluoroaniline 97%
5-(Benzyloxy)-2-fluoroaniline
Precaución: for Research Use Only


Formula: C₁₃H₁₂FNO
MW: 217.239 g/mol
MDL Number: MFCD00835609
CAS Number: 342042-82-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty