Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylthio)pyrazolo[1,5-a][1,3,5]triazin-4(3H)-one 95%

Supplier: AMBEED, INC
Warning

A133492-100MG A133492-250MG A133492-1G A133492-5G
77027-180EA 68 USD InStock
77027-180 77027-182 77666-922 77666-924
2-(Methylthio)pyrazolo[1,5-a][1,3,5]triazin-4(3H)-one 95%
2-(Methylthio)pyrazolo[1,5-a][1,3,5]triazin-4(3H)-one
Precaución: for Research Use Only


Formula: C₆H₆N₄OS
MDL Number: MFCD00661935
CAS Number: 54346-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty