Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(((tert-Butoxycarbonyl)(methyl)amino)methyl)phenyl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A134136-100MG A134136-1G A134136-250MG
77010-058EA 37 USD InStock
77010-058 77010-060 77010-062
(4-(((tert-Butoxycarbonyl)(methyl)amino)methyl)phenyl)boronic acid 98%
(4-(((tert-Butoxycarbonyl)(methyl)amino)methyl)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₃H₂₀BNO₄
MDL Number: MFCD11506067
CAS Number: 1287753-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty