Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-naphthol-6-sulfonate hydrate 98%

Supplier: AMBEED, INC
Warning

A134229-1000G A134229-100G A134229-500G A134229-25G
76996-564EA 87 USD InStock
76996-564 76996-566 76996-568 77298-698
Sodium-2-naphthol-6-sulfonate hydrate 98%
Sodium-2-naphthol-6-sulfonate hydrate
Precaución: for Research Use Only


Formula: C₁₀H₈O₄S
MW: 246.21 g/mol
MDL Number: MFCD00070488
CAS Number: 135-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty