Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol 95%

Supplier: AMBEED, INC
Warning

A134718-100G A134718-10G A134718-1G A134718-250MG A134718-25G A134718-5G
76983-914EA 198 USD InStock
76983-914 76983-916 76983-918 76983-920 76983-922 76983-924
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol 95%
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
Precaución: for Research Use Only


Formula: C₅H₃F₃N₂OS
MW: 196.10 g/mol
MDL Number: MFCD00112978
CAS Number: 368-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty