Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1-((tert-butoxycarbonyl)amino)-4-oxocyclohexanecarboxylate 98%

Supplier: AMBEED, INC
Warning

A134890-100MG A134890-250MG
76983-926EA 86 USD InStock
76983-926 76983-928
Methyl 1-((tert-butoxycarbonyl)amino)-4-oxocyclohexanecarboxylate 98%
Methyl 1-((tert-butoxycarbonyl)amino)-4-oxocyclohexanecarboxylate
Precaución: for Research Use Only


Formula: C₁₃H₂₁NO₅
MDL Number: MFCD13190094
CAS Number: 191111-27-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty