Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyanophenyl-trans-4-(4-n-propylcyclohexyl)benzoate 97%

Supplier: AMBEED, INC
Warning

A135485-10G A135485-1G A135485-5G
77027-282EA 34 USD InStock
77027-282 77027-284 77027-286
4-Cyanophenyl-trans-4-(4-n-propylcyclohexyl)benzoate 97%
4-Cyanophenyl-trans-4-(4-n-propylcyclohexyl)benzoate
Precaución: for Research Use Only


Formula: C₂₃H₂₅NO₂
MDL Number: MFCD09750943
CAS Number: 81930-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty