Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3S)-3-Phenylisoserine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A135538-100G A135538-10G A135538-1G A135538-25G A135538-5G
76996-612EA 288 USD InStock
76996-612 76996-614 76996-616 76996-618 76996-620
(2R,3S)-3-Phenylisoserine hydrochloride 97%
(2R,3S)-3-Phenylisoserine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClNO₃
MDL Number: MFCD09750971
CAS Number: 132201-32-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty