Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(9-Ethyl-9H-carbazol-3-yl)ethanone 98%

Supplier: AMBEED, INC
Warning

A135848-100MG A135848-1G A135848-250MG
77027-304EA 12 USD InStock
77027-304 77027-306 77027-308
1-(9-Ethyl-9H-carbazol-3-yl)ethanone 98%
1-(9-Ethyl-9H-carbazol-3-yl)ethanone
Precaución: for Research Use Only


Formula: C₁₆H₁₅NO
MDL Number: MFCD00814305
CAS Number: 1484-04-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty