Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-fluoro-4-methoxybenzoate 98%

Supplier: AMBEED, INC
Warning

A136344-1G A136344-250MG A136344-5G
76996-656EA 30 USD InStock
76996-656 76996-658 76996-660
Methyl 2-fluoro-4-methoxybenzoate 98%
Methyl 2-fluoro-4-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₉FO₃
MDL Number: MFCD06203758
CAS Number: 128272-26-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty