Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,6-dichloronicotinate 97%

Supplier: AMBEED, INC
Warning

A138568-100G A138568-1G A138568-250MG A138568-25G A138568-5G
76996-782EA 159 USD InStock
76996-782 76996-784 76996-786 76996-788 76996-790
Methyl-2,6-dichloronicotinate 97%
Methyl-2,6-dichloronicotinate
Precaución: for Research Use Only


Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD07369794
CAS Number: 65515-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty