Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxy-2-methylpyrimidin-4(3H)-one 97%

Supplier: AMBEED, INC
Warning

A138942-1000G A138942-100G A138942-10G A138942-25G A138942-500G A138942-5G
77010-270EA 193 USD InStock
77010-270 77010-272 77010-274 77010-276 77010-278 77010-280
6-Hydroxy-2-methylpyrimidin-4(3H)-one 97%
6-Hydroxy-2-methylpyrimidin-4(3H)-one
Precaución: for Research Use Only


Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD00205627
CAS Number: 1194-22-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty