Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tedizolid 98%

Supplier: AMBEED, INC
Danger

A139024-1G A139024-1MG A139024-250MG A139024-2MG A139024-5G A139024-5MG
76984-110EA 44 USD InStock
76984-110 76984-112 76984-114 76984-116 76984-118 76984-120
Tedizolid 98%
Tedizolid
Precaución: for Research Use Only


Formula: C₁₇H₁₅FN₆O₃
MW: 370.34 g/mol
MDL Number: MFCD19442562
CAS Number: 856866-72-3
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty