Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichlorobenzhydrazide 95%

Supplier: AMBEED, INC
Warning

A140021-10G A140021-1G A140021-250MG A140021-5G
77027-440EA 262 USD InStock
77027-440 77027-442 77027-444 77027-446
2,3-Dichlorobenzhydrazide 95%
2,3-Dichlorobenzhydrazide
Precaución: for Research Use Only


Formula: C₇H₆Cl₂N₂O
MDL Number: MFCD02612553
CAS Number: 438197-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty