Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dibromopyrazin-2-amine 97%

Supplier: AMBEED, INC
Warning

A140286-10G A140286-1G A140286-250MG A140286-5G
77027-484EA 207 USD InStock
77027-484 77027-486 77027-488 77027-490
3,6-Dibromopyrazin-2-amine 97%
3,6-Dibromopyrazin-2-amine
Precaución: For research use only.


Formula: C4H3Br2N3
MDL Number: MFCD11040377
CAS Number: 957230-70-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty