Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Ethyl-2-morpholinoethanamine 95%

Supplier: AMBEED, INC
Warning

A141037-10G A141037-1G A141037-250MG A141037-5G
76984-174EA 179 USD InStock
76984-174 76984-176 76984-178 76984-180
N-Ethyl-2-morpholinoethanamine 95%
N-Ethyl-2-morpholinoethanamine
Precaución: for Research Use Only


Formula: C₈H₁₈N₂O
MW: 158.24 g/mol
MDL Number: MFCD09802310
CAS Number: 108302-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty