Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate 98%

Supplier: AMBEED, INC
Warning

A141382-1G A141382-250MG A141382-25G A141382-5G
77027-556EA 48 USD InStock
77027-556 77027-558 77027-560 77027-562
tert-Butyl-4-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate 98%
tert-Butyl-4-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₆ClN₃O₂
MDL Number: MFCD08273922
CAS Number: 1056934-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty