Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide 95%

Supplier: AMBEED, INC
Warning

A141445-100MG A141445-1G A141445-250MG
76984-212EA 13 USD InStock
76984-212 76984-214 76984-216
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide 95%
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide
Precaución: for Research Use Only


Formula: C₁₄H₁₇Br₂NO₂S
MDL Number: MFCD23703120
CAS Number: 1231160-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty