Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methoxypyrimidine 96%

Supplier: AMBEED, INC
Warning

A142435-100G A142435-10G A142435-1G A142435-25G A142435-5G
77010-408EA 127 USD InStock
77010-408 77010-410 77010-412 77010-414 77010-416
4-Chloro-6-methoxypyrimidine 96%
4-Chloro-6-methoxypyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O
MW: 144.5590 g/mol
MDL Number: MFCD06200211
CAS Number: 26452-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty