Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK-1292263 98+%

Supplier: AMBEED, INC
Warning

A142714-100MG A142714-10MG A142714-1MG A142714-50MG A142714-5MG
76984-264EA 267 USD InStock
76984-264 76984-266 76984-268 76984-270 76984-272
GSK-1292263 98+%
GSK-1292263
Precaución: for Research Use Only


Formula: C₂₃H₂₈N₄O₄S
MDL Number: MFCD18385004
CAS Number: 1032823-75-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty