Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminoethanesulfonamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A143457-250MG A143457-25G
77010-462EA 15 USD InStock
77010-462 77010-464
2-Aminoethanesulfonamide hydrochloride 97%
2-Aminoethanesulfonamide hydrochloride
Precaución: for Research Use Only


Formula: C₂H₉ClN₂O₂S
MDL Number: MFCD02258341
CAS Number: 89756-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty