Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclopropyl-6,7-difluoro-8-hydroxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A143732-100MG A143732-1G A143732-250MG A143732-5G
76984-306EA 15 USD InStock
76984-306 76984-308 76984-310 76984-312
1-Cyclopropyl-6,7-difluoro-8-hydroxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 97%
1-Cyclopropyl-6,7-difluoro-8-hydroxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₉F₂NO₄
MDL Number: MFCD25973506
CAS Number: 154093-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty