Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A144246-10G A144246-1G A144246-250MG
77027-770EA 153 USD InStock
77027-770 77027-772 77027-774
2-Methyl-3-pyridineboronic acid pinacol ester 97%
2-Methyl-3-pyridineboronic acid pinacol ester


Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD08669594
CAS Number: 1012084-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty