Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dibromodibenzofuran 98%

Supplier: AMBEED, INC
Warning

A144917-100MG A144917-10G A144917-1G A144917-250MG A144917-25G A144917-5G
76984-402EA 5 USD InStock
76984-402 76984-404 76984-406 76984-408 76984-410 76984-412
4,6-Dibromodibenzofuran 98%
4,6-Dibromodibenzofuran
Precaución: for Research Use Only


Formula: C₁₂H₆Br₂O
MDL Number: MFCD00185686
CAS Number: 201138-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty