Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylbenzo[b]furan-2-carboxaldehyde 95%

Supplier: AMBEED, INC
Warning

A145667-1G A145667-250MG A145667-25G A145667-5G
76997-056EA 33 USD InStock
76997-056 76997-058 76997-060 76997-062
3-Methylbenzo[b]furan-2-carboxaldehyde 95%
3-Methylbenzo[b]furan-2-carboxaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₈O₂
MW: 160.169 g/mol
MDL Number: MFCD01459584
CAS Number: 1199-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty