Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-leucinol 97%

Supplier: AMBEED, INC
Warning

A145784-1G A145784-250MG A145784-5G A145784-25G
76997-078EA 19 USD InStock
76997-078 76997-080 76997-082 77671-070
Fmoc-L-leucinol 97%
Fmoc-L-leucinol
Precaución: for Research Use Only


Formula: C₂₁H₂₅NO₃
MDL Number: MFCD00235932
CAS Number: 139551-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty