Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-oxopentanedioate 95%

Supplier: AMBEED, INC
Warning

A145871-100MG A145871-1G A145871-250MG
77027-896EA 58 USD InStock
77027-896 77027-898 77027-900
Diethyl 2-oxopentanedioate 95%
Diethyl 2-oxopentanedioate
Precaución: for Research Use Only


Formula: C₉H₁₄O₅
MW: 202.20 g/mol
MDL Number: MFCD00032382
CAS Number: 5965-53-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty