Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Formylbis(triphenylphosphoranyl)rhodium(IV) chloride 98%

Supplier: AMBEED, INC
Warning

A146068-1G A146068-250MG
76997-084EA 135 USD InStock
76997-084 76997-086
Formylbis(triphenylphosphoranyl)rhodium(IV) chloride 98%
Formylbis(triphenylphosphoranyl)rhodium(IV) chloride
Precaución: for Research Use Only


Formula: C₃₇H₃₃ClOP₂Rh
MW: 693.96 g/mol
MDL Number: MFCD00044273
CAS Number: 13938-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty