Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-nitroisonicotinic acid 98%

Supplier: AMBEED, INC
Warning

A146478-100G A146478-10G A146478-1G A146478-250MG A146478-25G A146478-5G
77027-904EA 198 USD InStock
77027-904 77027-906 77027-908 77027-910 77027-912 77027-914
2-Chloro-5-nitroisonicotinic acid 98%
2-Chloro-5-nitroisonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃ClN₂O₄
MW: 202.552 g/mol
MDL Number: MFCD14584664
CAS Number: 907545-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty