Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-N-methylbenzamide 97%

Supplier: AMBEED, INC
Warning

A146506-10G A146506-1G A146506-25G A146506-5G
77027-916EA 53 USD InStock
77027-916 77027-918 77027-920 77027-922
4-Bromo-N-methylbenzamide 97%
4-Bromo-N-methylbenzamide
Precaución: for Research Use Only


Formula: C₈H₈BrNO
MDL Number: MFCD00462943
CAS Number: 27466-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty