Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorothiophenol 95%

Supplier: AMBEED, INC
Danger

A147270-1G A147270-250MG A147270-25G A147270-5G
77010-596EA 36 USD InStock
77010-596 77010-598 77010-600 77010-602
3,5-Difluorothiophenol 95%
3,5-Difluorothiophenol
Precaución: for Research Use Only


Formula: C₆H₄F₂S
MW: 146.16
MDL Number: MFCD06201795
CAS Number: 99389-26-1
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty