Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tripelennamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A147384-100G A147384-100MG A147384-1G A147384-250MG A147384-25G A147384-5G
77027-940EA 257 USD InStock
77027-940 77027-942 77027-944 77027-946 77027-948 77027-950
Tripelennamine hydrochloride 98%
Tripelennamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₆H₂₂ClN₃
MDL Number: MFCD00012815
CAS Number: 154-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty