Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-aminonicotinate 98%

Supplier: AMBEED, INC
Warning

A147875-100G A147875-10G A147875-1G A147875-25G A147875-500G A147875-5G
77010-626EA 99 USD InStock
77010-626 77010-628 77010-630 77010-632 77010-634 77010-636
Methyl-2-aminonicotinate 98%
Methyl-2-aminonicotinate
Precaución: for Research Use Only


Formula: C₇H₈N₂O₂
MW: 152.15
MDL Number: MFCD00128872
CAS Number: 14667-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty