Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Boc-2-ethylpiperazine 97%

Supplier: AMBEED, INC
Warning

A147913-10G A147913-1G A147913-5G A147913-100MG
76997-206EA 317 USD InStock
76997-206 76997-208 76997-210 77671-082
(S)-1-Boc-2-ethylpiperazine 97%
(S)-1-Boc-2-ethylpiperazine
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD07772100
CAS Number: 325145-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty