Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Bis(diphenylphosphino)hexane 97%

Supplier: AMBEED, INC
Warning

A148327-100G A148327-10G A148327-1G A148327-25G A148327-5G
76997-238EA 64 USD InStock
76997-238 76997-240 76997-242 76997-244 76997-246
1,6-Bis(diphenylphosphino)hexane 97%
1,6-Bis(diphenylphosphino)hexane
Precaución: for Research Use Only


Formula: C₃₀H₃₂P₂
MW: 454.53 g/mol
MDL Number: MFCD00003053
CAS Number: 19845-69-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty