Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride 98%

Supplier: AMBEED, INC
Warning

A148521-100G A148521-500G A148521-5G
76997-248EA 77 USD InStock
76997-248 76997-250 77671-088
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride 98%
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₈N₂O
MDL Number: MFCD03428357
CAS Number: 151257-01-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty