Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-(2,5-Difluorophenyl)propan-1-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A148844-100MG A148844-1G A148844-250MG
76984-566EA 56 USD InStock
76984-566 76984-568 76984-570
(S)-1-(2,5-Difluorophenyl)propan-1-amine hydrochloride 95%
(S)-1-(2,5-Difluorophenyl)propan-1-amine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClF₂N
MDL Number: MFCD12757712
CAS Number: 1391431-90-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty