Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropoxyaniline 97%

Supplier: AMBEED, INC
Warning

A149384-10G A149384-1G A149384-25G A149384-5G
76997-272EA 70 USD InStock
76997-272 76997-274 76997-276 76997-278
2-Isopropoxyaniline 97%
2-Isopropoxyaniline
Precaución: for Research Use Only


Formula: C₉H₁₃NO
MDL Number: MFCD00242947
CAS Number: 29026-74-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty