Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxocyclopent-1-enecarboxylic acid 95% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with hydroquinone (HQ) 0.1 %

A149781-100MG A149781-1G A149781-250MG
77028-064EA 133 USD InStock
77028-064 77028-066 77028-068
3-Oxocyclopent-1-enecarboxylic acid 95% stabilized
3-Oxocyclopent-1-enecarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₆O₃
MDL Number: MFCD11046428
CAS Number: 108384-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty