Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodo-2-methoxypyridine 98%

Supplier: AMBEED, INC
Warning

A149867-100G A149867-10G A149867-1G A149867-25G A149867-5G
76997-294EA 155 USD InStock
76997-294 76997-296 76997-298 76997-300 76997-302
3-Iodo-2-methoxypyridine 98%
3-Iodo-2-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₆INO
MDL Number: MFCD03094946
CAS Number: 112197-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty