Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-3-benzyloxy-1-propanol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A149893-1G A149893-250MG
76984-582EA 59 USD InStock
76984-582 76984-584
(R)-2-Amino-3-benzyloxy-1-propanol hydrochloride 95%
(R)-2-Amino-3-benzyloxy-1-propanol hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂·HCl
MW: 217.70 g/mol
MDL Number: MFCD04112476
CAS Number: 58577-95-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty