Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-bromobenzoyl formate 95%

Supplier: AMBEED, INC
Warning

A150012-100MG A150012-1G A150012-250MG A150012-25G A150012-5G
76997-304EA 6 USD InStock
76997-304 76997-306 76997-308 76997-310 76997-312
Ethyl-4-bromobenzoyl formate 95%
Ethyl-4-bromobenzoyl formate
Precaución: for Research Use Only


Formula: C₁₀H₉BrO₃
MDL Number: MFCD06659764
CAS Number: 20201-26-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty