Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-4-methyl-1H-imidazole 95%

Supplier: AMBEED, INC
Warning

A150783-10G A150783-1G A150783-250MG A150783-25G A150783-5G
76984-634EA 56 USD InStock
76984-634 76984-636 76984-638 76984-640 76984-642
5-Iodo-4-methyl-1H-imidazole 95%
5-Iodo-4-methyl-1H-imidazole
Precaución: for Research Use Only


Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD07783894
CAS Number: 15813-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty