Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-2-(2-methoxyethoxy)ethane 95%

Supplier: AMBEED, INC
Warning

A151382-100MG A151382-1G A151382-250MG A151382-5G
77010-796EA 15 USD InStock
77010-796 77010-798 77010-800 77010-802
1-Iodo-2-(2-methoxyethoxy)ethane 95%
1-Iodo-2-(2-methoxyethoxy)ethane
Precaución: for Research Use Only


Formula: C₅H₁₁IO₂
MDL Number: MFCD18205837
CAS Number: 104539-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty